Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

409340

Sigma-Aldrich

Samarium(II) iodide

anhydrous, powder, ≥99.9% trace metals basis

Synonym(s):

Samarium diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SmI2
CAS Number:
Molecular Weight:
404.17
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.9% trace metals basis
grade:
anhydrous
form:
powder

grade

anhydrous

Quality Level

Assay

≥99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: samarium

reagent type: reductant

impurities

≤1000.0 ppm Trace Rare Earth Analysis

SMILES string

I[Sm]I

InChI

1S/2HI.Sm/h2*1H;/q;;+2/p-2

InChI key

UAWABSHMGXMCRK-UHFFFAOYSA-L

Application

Promotes the cyanofluorination of aldehydes providing cyanofluorhydrins in good to excellent yield at room temperature in THF/HMPA.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, 1309-1309 (2007)
Martin Holan et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(27), 9877-9888 (2015-05-28)
Densely functionalized cyclopentane derivatives with up to four consecutive stereocenters are assembled by a tandem Michael addition/single-electron transfer oxidation/radical cyclization/oxygenation strategy mediated by ferrocenium hexafluorophosphate, a recyclable, less toxic single-electron transfer oxidant. Ester enolates were coupled with α-benzylidene and α-alkylidene

Articles

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Rare earth elements are vital in everyday life worldwide: catalysts in cars, colors in screens, magnets in electronics. Essential for modern living.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service