Skip to Content
MilliporeSigma
All Photos(1)

Documents

430633

Sigma-Aldrich

Aluminum L-lactate

95%

Synonym(s):

L-Lactic acid aluminum salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3CH(OH)COO]3Al
CAS Number:
Molecular Weight:
294.19
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

95%

form

solid

mp

>300 °C (lit.)

application(s)

battery manufacturing

SMILES string

C[C@H](O)C(=O)O[Al](OC(=O)[C@H](C)O)OC(=O)[C@H](C)O

InChI

1S/3C3H6O3.Al/c3*1-2(4)3(5)6;/h3*2,4H,1H3,(H,5,6);/q;;;+3/p-3/t3*2-;/m000./s1

InChI key

VXYADVIJALMOEQ-LGISMKCISA-K

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Elian-Simplice Yaganza et al.
Applied and environmental microbiology, 75(5), 1465-1469 (2008-12-31)
Twenty-one salts were tested for their effects on the growth of Pectobacterium carotovorum subsp. carotovorum and Pectobacterium atrosepticum. In liquid medium, 11 salts (0.2 M) exhibited strong inhibition of bacterial growth. The inhibitory action of salts relates to the water-ionizing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service