MilliporeSigma
  • Physicochemical basis for the inhibitory effects of organic and inorganic salts on the growth of Pectobacterium carotovorum subsp. carotovorum and Pectobacterium atrosepticum.
Physicochemical basis for the inhibitory effects of organic and inorganic salts on the growth of Pectobacterium carotovorum subsp. carotovorum and Pectobacterium atrosepticum.
Applied and environmental microbiology (2008-12-31)
Elian-Simplice Yaganza, Russell J Tweddell, Joseph Arul
ABSTRACT
Twenty-one salts were tested for their effects on the growth of Pectobacterium carotovorum subsp. carotovorum and Pectobacterium atrosepticum. In liquid medium, 11 salts (0.2 M) exhibited strong inhibition of bacterial growth. The inhibitory action of salts relates to the water-ionizing capacity and the lipophilicity of their constituent ions.
Sigma-Aldrich
1-Octanol, JIS special grade, ≥98.0%
Sigma-Aldrich
1-Octanol, SAJ first grade, ≥75.0%
Sigma-Aldrich
1-Octanol, natural, ≥98%, FCC
Sigma-Aldrich
1-Octanol, ReagentPlus®, 99%
Supelco
1-Octanol, analytical standard
Sigma-Aldrich
1-Octanol, ≥98%, FCC, FG
Sigma-Aldrich
1-Octanol, suitable for HPLC, ≥99%
Sigma-Aldrich
1-Octanol, ACS reagent, ≥99%
Sigma-Aldrich
1-Octanol, anhydrous, ≥99%
Sigma-Aldrich
Aluminum L-lactate, 95%