Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

294853

Sigma-Aldrich

Aluminum acetate, dibasic

contains boric acid as stabilizer

Synonym(s):

Dihydroxyaluminum acetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5O4Al
CAS Number:
Molecular Weight:
120.04
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

contains

boric acid as stabilizer

composition

Al, 16-19% EDTA titration
B, 2.5% EDTA titration

reaction suitability

core: aluminum

SMILES string

CC(=O)O[Al](O)O

InChI

1S/C2H4O2.Al.2H2O/c1-2(3)4;;;/h1H3,(H,3,4);;2*1H2/q;+3;;/p-3

InChI key

KLMDYFUUSKOJAX-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

General description

Aluminum acetate dibasic is a white powder that has been stabilized with boric acid. It is a water-soluble compound with the ability to hydrolyze in aqueous solutions. Additionally, it has been investigated for its potential as a corrosion inhibitor due to its properties as a Lewis acid.

Application

Aluminum acetate, dibasic can be used as:
  • As an aluminum dopant to reduce the cobalt content in lithium-rich layered oxide cathode materials of lithium-ion batteries. These materials exhibit excellent capacity retention and voltage retention, enhancing electrochemical performance.
  • To prepare doped zinc oxide thin films by atmospheric pressure plasma (APP) deposition process.
  • As a cross-linker for temperature-controlled gel-casting and joining of ceramics. These aluminum borate nanofibrous porous ceramics are excellent candidates for high-temperature insulation applications.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Aluminium acetate as alternative cross-linker for temperature-controlled gel-casting and joining of ceramics
Christoph Brandes, et al.
J. Eur. Ceram. Soc., 5, 1241-1251 (2016)
Aluminium acetate as alternative cross-linker for temperature-controlled gel-casting and joining of ceramics
Christoph Brandes, et al
J. Eur. Ceram. Soc., 36, 1241-1251 (2016)
Aluminum doped zinc oxide coatings at low temperature by atmospheric pressure plasma jet
M. Favaro, et al.
Thin Solid Films, 708, 13811-13811 (2020)
Impact of Overlithiation and Al doping on the battery performance of Li-rich layered oxide materials
A. Celeste, et al.
Electrochimica Acta, 428, 140737-140737 (2022)

Articles

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

The diversity of applications and nanostructured materials accessible using ultrasonic spray methods are highlighted in this article.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service