Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

422908

Sigma-Aldrich

2-Methyl-4-nitrophenol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)OH
CAS Number:
Molecular Weight:
153.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

93-98 °C (lit.)

functional group

nitro

SMILES string

Cc1cc(ccc1O)[N+]([O-])=O

InChI

1S/C7H7NO3/c1-5-4-6(8(10)11)2-3-7(5)9/h2-4,9H,1H3

InChI key

KDQPMQNHVQVVMR-UHFFFAOYSA-N

General description

2-Methyl-4-nitrophenol is a nitrophenol derivative. The study of its crystal structure reveals the molecule to be nearly planar. Its synthesis has been reported. FT-IR and FT-Raman spectra and molecular modeling of 2-methyl-4-nitrophenol have been analyzed. It is reported to be one of the compounds in diesel exhaust particles (DEP) that shows potential vasodilatation activity in rats. Its reduction catalyzed by porous carbon-encapsulated gold nanoparticles has been investigated.

Application

2-Methyl-4-nitrophenol is suitable as a target compound in the study on measurement of methylnitrophenol concentrations and stable isotope ratios in the atmospheric particulate matter and as an internal standard in the determination of monoaromatic nitro compounds in atmospheric aerosols using high performance liquid chromatography electrospray tandem mass spectrometry (HPLC/ESI-MS/MS) method.
It may be used as starting material in the synthesis of 2-bromo-4-nitro-6-methylphenol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of Photosynthesis by 4-Nitro-6-alkyl-phenols: Structure-Activity Studies in Wild Type and Five Mutants of Chlamydomonas reinhardtii Thylakoids.
Draber W, et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 48, 213-213 (1993)
Jinrui Guo et al.
Chemical communications (Cambridge, England), 48(90), 11094-11096 (2012-10-11)
Gold nanoparticles encapsulated in porous carbon spheres with high surface areas (~800 m(2) g(-1)) have been synthesized by ultrasonic spray pyrolysis (USP). The porous carbon-encapsulated gold nanoparticles exhibit excellent catalytic activity toward the reductions of both hydrophilic and hydrophobic nitroaromatics.
2-Methyl-4-nitrophenol.
Bi S, et al.
Acta Crystallographica Section E, Structure Reports Online, 65(6), 1378-1378 (2009)
Zoran Kitanovski et al.
Journal of chromatography. A, 1268, 35-43 (2012-11-06)
Nitrogen-containing organic compounds in the atmosphere have drawn attention owing to their impact on aerosol chemistry and physics and their potential adverse effects on the biosphere. Among them, nitrocatechols and their homologs have recently been associated with biomass burning. In
Yoki Mori et al.
Biological & pharmaceutical bulletin, 26(3), 394-395 (2003-03-04)
The compounds in diesel exhaust particles (DEP) that are responsible for vasodilatation were isolated and characterized for the first time. From benzene extract of DEP, 2-methyl-4-nitrophenol, 3-methyl-4-nitrophenl and 4-nitrophenol were isolated, and their vasodilatation activities were confirmed. 3-methyl-4-nitrophenol caused dilatation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service