Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

288659

Sigma-Aldrich

Iron(III) fluoride

46.5-50.7% Fe (by Na2SO3, titration)

Synonym(s):

Ferric fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FeF3
CAS Number:
Molecular Weight:
112.84
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

form

powder

concentration

46.5-50.7% Fe (by Na2SO3, titration)

application(s)

battery manufacturing

SMILES string

F[Fe](F)F

InChI

1S/3FH.Fe/h3*1H;/q;;;+3/p-3

InChI key

SHXXPRJOPFJRHA-UHFFFAOYSA-K

General description

Iron(III) fluoride, also known as ferric fluoride or iron trifluoride. It is a thermally stable inorganic compound widely used as a cathode material for lithium-ion batteries due to its high capacity, relatively high working potential, and reversibility compared to other transition metal fluorides. Additionally, it is also used as a catalyst in certain organic reactions, such as the chemoselective addition of cyanide to aldehydes.

Application

Iron(III) fluoride can be used:      
  • To produce high-capacity lithium storing cathodes via liquid phase exfoliation method.     
  • As a cathode material for sodium-ion batteries.     
  • As a catalyst for the conversion of carbonyl compounds into oxathioacetals and dithioacetals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A review on the application of iron (III) fluorides as positive electrodes for secondary cells
Donato E Conte, et al.},
Materials for renewable and sustainable energy, 3, 1-22 (2014)
Iron (III) fluoride: A highly efficient and versatile catalyst for the protection of carbonyl compounds under solvent-free conditions
Babasaheb P Bandgar, et al.
Australian Journal of Chemistry, 58, 607-610 (2005)

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service