Skip to Content
MilliporeSigma
All Photos(1)

Documents

137790

Sigma-Aldrich

5-Methyl-2-nitrophenol

97%

Synonym(s):

3-Hydroxy-4-nitrotoluene, 6-Nitro-m-cresol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)OH
CAS Number:
Molecular Weight:
153.14
Beilstein:
1868030
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

53-56 °C (lit.)

SMILES string

Cc1ccc(c(O)c1)[N+]([O-])=O

InChI

1S/C7H7NO3/c1-5-2-3-6(8(10)11)7(9)4-5/h2-4,9H,1H3

InChI key

NQXUSSVLFOBRSE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Methyl-2-nitrophenol is a chlorination by-product of butamifos.

Application

5-Methyl-2-nitrophenol was used in the synthesis of 3-methoxy-4-nitrotoluene.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

228.2 °F

Flash Point(C)

109 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R I Olariu et al.
Environmental science & technology, 47(14), 7729-7738 (2013-06-12)
Product analyses of the NO3 radical-initiated oxidation of ortho-, meta-, and para-cresol have been performed in large-volume chamber systems at the University of Wuppertal (1080 L quartz glass reactor: QUAREC) and the European Photoreactor (EUPHORE), Valencia, Spain. The reaction of
Hideki Onagi et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 9(24), 5971-5977 (2003-12-18)
The conformations of two [2]rotaxanes, each comprising alpha-cyclodextrin as the rotor, a stilbene as the axle and 2,4,6-trinitrophenyl substituents as the capping groups, have been examined in solution and in the solid state, using (1)H NMR spectroscopy and X-ray crystallography
J I Cacho et al.
Journal of chromatography. A, 1456, 27-33 (2016-06-19)
A rapid and sensitive procedure for the determination of six NPs in soils by gas chromatography and mass spectrometry (GC-MS) is proposed. Ultrasound assisted extraction (UAE) is used for NP extraction from soil matrices to an organic solvent, while the
Marta Pastor-Belda et al.
Talanta, 189, 543-549 (2018-08-09)
This paper presents a procedure for the determination of seven nitrophenols (NPs) in water and soil samples using stir bar sorptive extraction (SBSE) coupled to gas chromatography with mass spectrometry (GC-MS) by means of a thermal desorption unit (TDU). Microwave
Masahiro Kamoshita et al.
Chemosphere, 78(4), 482-487 (2009-10-30)
The mutagenic activities of 5-methyl-2-nitrophenol (5M2NP), a chlorination by-product of butamifos, its structural isomer 2-methyl-5-nitrophenol (2M5NP), and related compounds were evaluated by the Ames assay. The mutagenic activities of 5M2NP and 2M5NP were negative or not particularly high. However, those

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service