Skip to Content
MilliporeSigma
All Photos(1)

Documents

422169

Sigma-Aldrich

2,6-Dichloro-5-fluoro-3-pyridinecarbonitrile

98%

Synonym(s):

2,6-Dichloro-3-cyano-5-fluoropyridine, 2,6-Dichloro-5-fluoro-3-nicotinonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6HCl2FN2
CAS Number:
Molecular Weight:
190.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

mp

89-91 °C (lit.)

SMILES string

Fc1cc(C#N)c(Cl)nc1Cl

InChI

1S/C6HCl2FN2/c7-5-3(2-10)1-4(9)6(8)11-5/h1H

InChI key

DEDKKOOGYIMMBC-UHFFFAOYSA-N

General description

2,6-Dichloro-5-fluoro-3-pyridinecarbonitrile is a pyridine derivative. It is a fluorinated heterocyclic building block used in chemical synthesis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service