Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

736066

Sigma-Aldrich

3,5-Difluoropyridine-2-carbonitrile

97%

Synonym(s):

2-Cyano-3,5-difluorpyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2F2N2
CAS Number:
Molecular Weight:
140.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

30-34 °C

SMILES string

Fc1cnc(C#N)c(F)c1

InChI

1S/C6H2F2N2/c7-4-1-5(8)6(2-9)10-3-4/h1,3H

InChI key

WLBIFECTHKFYKV-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

194.0 °F

Flash Point(C)

90 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service