Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

303526

Sigma-Aldrich

2-Amino-5-bromopyrimidine

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4BrN3
CAS Number:
Molecular Weight:
174.00
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

241-243 °C (lit.)

SMILES string

Nc1ncc(Br)cn1

InChI

1S/C4H4BrN3/c5-3-1-7-4(6)8-2-3/h1-2H,(H2,6,7,8)

InChI key

UHRHPPKWXSNZLR-UHFFFAOYSA-N

General description

Crystal structure of 2-amino-5-bromopyrimidine was studied.

Application

2-Amino-5-bromopyrimidine was employed:
  • in synthesis of pyridine, pyrimidine and pyridinone C-nucleoside phosphoramidites
  • in synthesis of 2-phthalimido-5-bromopyrimidine
  • as intermediate for the preparation of sulfanilamides and amino acids containing the pyrimidine ring system. The products are potential antiviral agents

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure of 2-amino-5-bromopyrimidine.
Watton HLL, et al.
Acta Crystallographica Section C, Crystal Structure Communications, 44(10), 1857-1858 (1988)
Journal of Chemical Education, 62, 905-905 (1985)
Jun Lu et al.
The Journal of organic chemistry, 74(21), 8021-8030 (2009-10-02)
In the structures of the HDV ribozyme a cytosine nucleobase resides at the active site poised to participate directly in catalysis. Defining the functional role of the nucleobase requires nucleoside analogues that perturb the functional groups in a strategic manner.
Self-assembly of 1-and 2-dimensional multicompartmental arrays via the 2-aminopyrimidine H-bonding motif and selective guest inclusion.
Krische MJ, et al.
Tetrahedron, 56(36), 6701-6706 (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service