Skip to Content
MilliporeSigma
All Photos(3)

Documents

412406

Sigma-Aldrich

PAMAM dendrimer, ethylenediamine core, generation 2.0 solution

20 wt. % in methanol

Synonym(s):

PAMAM, PAMAM-G 2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=2);dendri PAMAM(NH2)16
CAS Number:
Molecular Weight:
3256.18
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23

description

amino surface groups

Quality Level

form

liquid

mol wt

generation 2

feature

core type ethylenediamine core (2-carbon core)

concentration

20 wt. % in methanol

no. Surface Groups

16

refractive index

n20/D 1.362

bp

64.7 °C

density

0.86 g/mL at 25 °C

storage temp.

2-8°C

InChI

1S/C142H288N58O28/c143-29-45-159-115(201)1-73-187(74-2-116(202)160-46-30-144)101-61-175-131(217)17-89-195(90-18-132(218)176-62-102-188(75-3-117(203)161-47-31-145)76-4-118(204)162-48-32-146)109-69-183-139(225)25-97-199(98-26-140(226)184-70-110-196(91-19-133(219)177-63-103-189(77-5-119(205)163-49-33-147)78-6-120(206)164-50-34-148)92-20-134(220)178-64-104-190(79-7-121(207)165-51-35-149)80-8-122(208)166-52-36-150)113-114-200(99-27-141(227)185-71-111-197(93-21-135(221)179-65-105-191(81-9-123(209)167-53-37-151)82-10-124(210)168-54-38-152)94-22-136(222)180-66-106-192(83-11-125(211)169-55-39-153)84-12-126(212)170-56-40-154)100-28-142(228)186-72-112-198(95-23-137(223)181-67-107-193(85-13-127(213)171-57-41-155)86-14-128(214)172-58-42-156)96-24-138(224)182-68-108-194(87-15-129(215)173-59-43-157)88-16-130(216)174-60-44-158/h1-114,143-158H2,(H,159,201)(H,160,202)(H,161,203)(H,162,204)(H,163,205)(H,164,206)(H,165,207)(H,166,208)(H,167,209)(H,168,210)(H,169,211)(H,170,212)(H,171,213)(H,172,214)(H,173,215)(H,174,216)(H,175,217)(H,176,218)(H,177,219)(H,178,220)(H,179,221)(H,180,222)(H,181,223)(H,182,224)(H,183,225)(H,184,226)(H,185,227)(H,186,228)

InChI key

QYMWEDAKQRTCGD-UHFFFAOYSA-N

Application

Used in MRI contrast agents. Used in the condensation formation of silica nanospheres.

Physical form

Contains 16 surface primary amino groups

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc R Knecht et al.
Langmuir : the ACS journal of surfaces and colloids, 20(11), 4728-4732 (2005-06-23)
In diatoms, silica synthesis occurs by use of complex posttranslationally modified peptides, termed silaffins, and highly complex biological polyamine structures. Silaffin peptides have lysine residues that are modified to long-chain polyamine moieties of N-methyl derivatives of polypropylenimine to drive silica
Kobayashi, H., et al.
Magn. Reson. in Med., 50, 758-758 (2003)

Articles

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service