Skip to Content
MilliporeSigma
All Photos(1)

Documents

412376

Sigma-Aldrich

PAMAM dendrimer, ethylenediamine core, generation 0.5 solution

20 wt. % in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=0.5);dendri PAMAM(NHCH2CH2COONa)8
Molecular Weight:
1269.04
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

description

sodium carboxylate surface groups

Quality Level

form

liquid

mol wt

generation 0.5

feature

core type ethylenediamine core (2-carbon core)

concentration

20 wt. % in methanol

no. Surface Groups

8

refractive index

n20/D 1.368

bp

65 °C

density

0.86 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[O-]C(=O)CCN(CCNC(=O)CCN(CCN(CCC(=O)NCCN(CCC([O-])=O)CCC([O-])=O)CCC(=O)NCCN(CCC([O-])=O)CCC([O-])=O)CCC(=O)NCCN(CCC([O-])=O)CCC([O-])=O)CCC([O-])=O

InChI

1S/C46H80N10O20.8Na/c57-35(47-13-29-51(21-5-39(61)62)22-6-40(63)64)1-17-55(18-2-36(58)48-14-30-52(23-7-41(65)66)24-8-42(67)68)33-34-56(19-3-37(59)49-15-31-53(25-9-43(69)70)26-10-44(71)72)20-4-38(60)50-16-32-54(27-11-45(73)74)28-12-46(75)76;;;;;;;;/h1-34H2,(H,47,57)(H,48,58)(H,49,59)(H,50,60)(H,61,62)(H,63,64)(H,65,66)(H,67,68)(H,69,70)(H,71,72)(H,73,74)(H,75,76);;;;;;;;/q;8*+1/p-8

InChI key

HLBBXYHSZWSLRS-UHFFFAOYSA-F

Physical form

Contains 8 surface carboxylate groups

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service