Skip to Content
MilliporeSigma
All Photos(1)

Documents

411523

Sigma-Aldrich

3,5-Dibromobenzoic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2C6H3CO2H
CAS Number:
Molecular Weight:
279.91
Beilstein:
1940691
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

mp

218-220 °C (lit.)

SMILES string

OC(=O)c1cc(Br)cc(Br)c1

InChI

1S/C7H4Br2O2/c8-5-1-4(7(10)11)2-6(9)3-5/h1-3H,(H,10,11)

InChI key

SFTFNJZWZHASAQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,5-Dibromobenzoic acid is 3,5-dibromo-substituted benzoic acid.

Application

3,5-Dibromobenzoic acid may be used for the following syntheses:
  • (+)-menthyl 3,5-dibromobenzoate
  • di-tert-butyl 4-[2-(tert-butoxycarbonyl)ethyl]-4-(3,5-dibromobenzamido)heptanedioate
  • (L)-methyl 2-(3,5-dibromobenzamido)-3-phenylpropanoate

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Syntheses and enantioselective recognition of chiral poly (phenyleneethynylene) s bearing bulky optically active menthyl groups
Shinohara K-i, et al.
Polymer, 42(1), 351-355 (2001)
Synthesis of 5-substituted 1, 3-[bis (2, 2': 6', 2 ?-terpyridin-4'-ylethynyl)] benzene ligands and their coordination-driven self-assembly.
Hwang, S-H, et al.
Designed Monomers and Polymers, 9(5), 413-424 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service