Skip to Content
MilliporeSigma
All Photos(2)

Documents

646105

Sigma-Aldrich

3,5-Dibromophenol

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4Br2O
CAS Number:
Molecular Weight:
251.90
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

solid

mp

79-83 °C (lit.)

SMILES string

Oc1cc(Br)cc(Br)c1

InChI

1S/C6H4Br2O/c7-4-1-5(8)3-6(9)2-4/h1-3,9H

InChI key

PZFMWYNHJFZBPO-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shengnan You et al.
Soft matter, 13(18), 3334-3340 (2017-04-20)
Rod-coil molecules 1a, 1b and 2a, 2b, consisting of biphenyl and phenyl units connected by an acetylene bond as the rod segment and oligo(ethylene glycol) (OEG) as the coil segment, were synthesized and characterized. Molecules 1a and 1b incorporate a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service