Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

400378

Sigma-Aldrich

Lithium phenoxide solution

1.0 M in THF

Synonym(s):

Phenol lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5OLi
CAS Number:
Molecular Weight:
100.04
Beilstein:
3569793
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

reaction suitability

core: lithium

concentration

1.0 M in THF

bp

67 °C

density

0.918 g/mL at 25 °C

SMILES string

[Li+].[O-]c1ccccc1

InChI

1S/C6H6O.Li/c7-6-4-2-1-3-5-6;/h1-5,7H;/q;+1/p-1

InChI key

XAVQZBGEXVFCJI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-2.2 °F - closed cup

Flash Point(C)

-19 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jean-Pierre Brog et al.
Journal of nanobiotechnology, 15(1), 58-58 (2017-08-24)
LiCoO Several new single source precursors containing lithium (Li Our heterobimetallic single source precursors allow to dramatically reduce the production temperature and time for HT-LCO. The obtained nanoparticles of LiCoO

Articles

Few Monolayer Atomic Layer Deposition (ALD) on Surfaces and Interfaces for Energy Applications

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service