Skip to Content
MilliporeSigma
All Photos(1)

Documents

400246

Sigma-Aldrich

Lithium ethoxide solution

1.0 M in ethanol

Synonym(s):

Lithium ethanolate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OLi
CAS Number:
Molecular Weight:
52.00
Beilstein:
3909950
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

reaction suitability

core: lithium

concentration

1.0 M in ethanol

density

0.821 g/mL at 25 °C

SMILES string

[Li+].CC[O-]

InChI

1S/C2H5O.Li/c1-2-3;/h2H2,1H3;/q-1;+1

InChI key

AZVCGYPLLBEUNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Self-heat. 1 - Skin Corr. 1B

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

48.2 °F - closed cup

Flash Point(C)

9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service