Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

400270

Sigma-Aldrich

Lithium thiophenolate solution

1.0 M in THF

Synonym(s):

Thiophenol lithium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5SLi
CAS Number:
Molecular Weight:
116.11
Beilstein:
3597301
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

Quality Level

reaction suitability

core: lithium

concentration

1.0 M in THF

bp

67 °C

density

0.934 g/mL at 25 °C

SMILES string

[Li].Sc1ccccc1

InChI

1S/C6H6S.Li/c7-6-4-2-1-3-5-6;/h1-5,7H;/q;+1/p-1

InChI key

HPFQTCRYSOTMDJ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Few Monolayer Atomic Layer Deposition (ALD) on Surfaces and Interfaces for Energy Applications

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service