Skip to Content
MilliporeSigma
All Photos(2)

Documents

374725

Sigma-Aldrich

Lithium oxide

97%, powder, −60 mesh

Synonym(s):

Dilithium monoxide, Dilithium oxide, Kickerite, Lithia

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Li2O
CAS Number:
Molecular Weight:
29.88
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

powder

particle size

−60 mesh

density

2.013 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

[Li+].[Li+].[O--]

InChI

1S/2Li.O/q2*+1;-2

InChI key

FUJCRWPEOMXPAD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Lithium oxide is a white crystalline solid. Its a strong base. It reacts with water and forms lithium hydroxide.

Application

  • A review of high-energy density lithium-air battery technology: Investigates the effect of oxides and nanocatalysts on lithium oxide placement in batteries, offering insights into advanced battery technologies (Suryatna et al., 2022).
  • Dry-air-stable lithium silicide–lithium oxide core–shell nanoparticles: Discusses high-capacity prelithiation reagents for lithium-ion batteries, highlighting the role of lithium oxide in enhancing battery performance (Zhao et al., 2014).
  • A high-energy-density lithium-oxygen battery based on a reversible four-electron conversion to lithium oxide: Explores a novel battery chemistry that involves lithium oxide, emphasizing its potential to significantly increase energy density (Xia et al., 2018).
  • Lithium oxide superionic conductors inspired by garnet and NASICON structures: This study focuses on lithium oxide-based conductors for battery applications, demonstrating the potential of lithium oxide in solid-state battery technologies (Xiao et al., 2021).
  • Solution‐processed alkaline lithium oxide dielectrics for applications in n‐and p‐type thin‐film transistors: Reviews the use of lithium oxide in electronic applications, particularly for enhancing the performance of thin-film transistors (Liu et al., 2016).

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service