Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

381675

Sigma-Aldrich

Dytek® EP diamine

98%

Synonym(s):

1,3-Diaminopentane, 1,3-Pentanediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(NH2)CH2CH2NH2
CAS Number:
Molecular Weight:
102.18
Beilstein:
6796726
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

refractive index

n20/D 1.452 (lit.)

bp

164 °C (lit.)

mp

−121 °C (lit.)

density

0.855 g/mL at 25 °C (lit.)

SMILES string

CCC(N)CCN

InChI

1S/C5H14N2/c1-2-5(7)3-4-6/h5H,2-4,6-7H2,1H3

InChI key

WTSXICLFTPPDTL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

DYTEK is a registered trademark of Invista North America S.a.r.l.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephany Y Tzeng et al.
International journal of nanomedicine, 6, 3309-3322 (2012-01-10)
Biodegradable poly(ester amine) (PEA)-based and poly(amido amine) (PAA)-based nanoparticles were developed for efficient in vitro siRNA delivery to human umbilical vein endothelial cells (HUVECs). They were screened, characterized, and compared with traditionally studied DNA-containing particles. Several of the polymeric nanoparticles
Camila G Zamboni et al.
Journal of controlled release : official journal of the Controlled Release Society, 263, 18-28 (2017-03-30)
Hepatocellular carcinoma (HCC) is the third most deadly cancer in the US, with a meager 5-year survival rate of <20%. Such unfavorable numbers are closely related to the heterogeneity of the disease and the unsatisfactory therapies currently used to manage
Antonella Mangraviti et al.
ACS nano, 9(2), 1236-1249 (2015-02-03)
Biodegradable polymeric nanoparticles have the potential to be safer alternatives to viruses for gene delivery; however, their use has been limited by poor efficacy in vivo. In this work, we synthesize and characterize polymeric gene delivery nanoparticles and evaluate their
Jieun Hong et al.
Journal of controlled release : official journal of the Controlled Release Society, 200, 212-221 (2015-01-03)
Lineage conversion from one somatic cell type to another is an attractive approach for deriving specific therapeutic cell generation. In order to bypass inducing pluripotent stage, transdifferentiation/direct conversion technologies have been recently developed. We report the development of a direct
Feng Zhu et al.
ACS nano, 9(3), 2948-2959 (2015-02-11)
Organometallic halide perovskites CH3NH3PbX3 (X = I, Br, Cl) have quickly become one of the most promising semiconductors for solar cells, with photovoltaics made of these materials reaching power conversion efficiencies of near 20%. Improving our ability to harness the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service