Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

329665

Sigma-Aldrich

1,5-Diamino-2-methylpentane

99%

Synonym(s):

2-Methyl-1,5-pentanediamine, 2-Methylpentamethylene diamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2CH2CH(CH3)CH2NH2
CAS Number:
Molecular Weight:
116.20
Beilstein:
1732701
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

130 mmHg ( 135 °C)

Assay

99%

form

liquid

autoignition temp.

568 °F

refractive index

n20/D 1.459 (lit.)

bp

193 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

SMILES string

CC(CN)CCCN

InChI

1S/C6H16N2/c1-6(5-8)3-2-4-7/h6H,2-5,7-8H2,1H3

InChI key

JZUHIOJYCPIVLQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

181.4 °F - closed cup

Flash Point(C)

83 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jayoung Kim et al.
Acta biomaterialia, 41, 293-301 (2016-06-06)
The design of polymeric nanoparticles for gene therapy requires engineering of polymer structure to overcome multiple barriers, including prolonged colloidal stability during formulation and application. Poly(β-amino ester)s (PBAEs) have been shown effective as polymeric vectors for intracellular DNA delivery, but
Camila G Zamboni et al.
Journal of controlled release : official journal of the Controlled Release Society, 263, 18-28 (2017-03-30)
Hepatocellular carcinoma (HCC) is the third most deadly cancer in the US, with a meager 5-year survival rate of <20%. Such unfavorable numbers are closely related to the heterogeneity of the disease and the unsatisfactory therapies currently used to manage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service