Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

377163

Sigma-Aldrich

Tetrakis(acetonitrile)silver(I) tetrafluoroborate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3CN)4Ag]BF4
CAS Number:
Molecular Weight:
358.88
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

reaction suitability

core: silver
reagent type: catalyst

mp

72-75 °C (lit.)

SMILES string

[Ag+].CC#N.CC#N.CC#N.CC#N.F[B-](F)(F)F

InChI

1S/4C2H3N.Ag.BF4/c4*1-2-3;;2-1(3,4)5/h4*1H3;;/q;;;;+1;-1

InChI key

XZYAKBATRMXJSW-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Luminescent face-to-face dinuclear platinum(II) alkynyl phosphine silver(I) complex
  • Alpha-diimine silver(I) complexes
  • Silver(I) complexes with multidentate pyrazolyl-pyridine ligands
  • Silver(I) clusters with binding for a dithiophosphate ligand

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service