Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

333875

Sigma-Aldrich

Zinc tetrafluoroborate hydrate

Synonym(s):

Zinc ditetrafluoroborate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Zn(BF4)2·xH2O
CAS Number:
Molecular Weight:
239.00 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
crystalline

form

crystalline

composition

Degree of hydration, 6-7

reaction suitability

reagent type: catalyst
core: zinc

SMILES string

O.[Zn++].F[B-](F)(F)F.F[B-](F)(F)F

InChI

1S/2BF4.H2O.Zn/c2*2-1(3,4)5;;/h;;1H2;/q2*-1;;+2

InChI key

WKVXCUNBQMAPGE-UHFFFAOYSA-N

Application

Starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properites.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy C. Higgs et al.
Inorganic chemistry, 37(9), 2263-2272 (2001-10-24)
The mixed functionality pyrazole/phenol ligand (2-hydroxyphenyl)bis(pyrazolyl)methane, L1OH, has been used to prepare a series of linear trimetallic systems with the general structural motif [M(3)(L1O)(4)](2+), where M = Mn(2+), Co(2+), Ni(2+), Cu(2+), and Zn(2+). Each of these complexes has been structurally

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service