Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

572144

Sigma-Aldrich

Bromotris(triphenylphosphine)copper(I)

98%

Synonym(s):

Bromotris(triphenylphosphine)copper, Tris(triphenylphosphine)copper(I) bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C54H45BrP3Cu
CAS Number:
Molecular Weight:
930.31
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

solid

reaction suitability

reagent type: catalyst
core: copper

mp

168-170 °C (lit.)

SMILES string

[Cu]Br.c1ccc(cc1)P(c2ccccc2)c3ccccc3.c4ccc(cc4)P(c5ccccc5)c6ccccc6.c7ccc(cc7)P(c8ccccc8)c9ccccc9

InChI

1S/3C18H15P.BrH.Cu/c3*1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;;/h3*1-15H;1H;/q;;;;+1/p-1

InChI key

QXLPGKOIQZWUPL-UHFFFAOYSA-M

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gurjaspreet Singh et al.
European journal of medicinal chemistry, 108, 287-300 (2015-12-24)
A series of chalconyl blended triazole allied silatranes (7a-g/8a-g/9a-g) were synthesized in good yields using a simple, economical and biocompatible synthetic route. The blend of three different pharmacologically active moieties into a single scaffold resulted into synergistic effect in their

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service