336637
Lithium aluminate
Synonym(s):
Aluminum lithium oxide
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
LiAlO2
CAS Number:
Molecular Weight:
65.92
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
form
powder and chunks
density
2.615 g/mL at 25 °C (lit.)
SMILES string
[Li+].[O-][Al]=O
InChI
1S/Al.Li.2O/q;+1;;-1
InChI key
YQNQTEBHHUSESQ-UHFFFAOYSA-N
Related Categories
General description
Lithium aluminate (LiAlO₂) is a versatile inorganic compound with high thermal stability, low density, and excellent ion conductivity. It is suitable for a range of applications, such as radiation dosimetry, where it serves as an effective optically stimulated luminescence(OSL) dosimeter, and in nuclear technology as a tritium breeding material for fusion reactors. Additionally, it is widely used in development of solid polymer electrolytes for batteries, enhancing conductivity and performance in energy storage systems.
Application
- Lithium‐Aluminate‐Catalyzed Hydrophosphination Applications: This study probes the catalytic potential of lithium aluminates in hydrophosphination reactions, specifically utilizing [iBu3AlHLi]2 (VA Pollard, A Young, R McLellan, 2019).
- Tailoring Lithium Aluminate Phases Based on Thermodynamics for an Increased Recycling Efficiency of Li-Ion Batteries: This paper discusses the thermodynamic tailoring of lithium aluminate phases to enhance the recycling efficiency of lithium-ion batteries (H Li, H Qiu, T Schirmer, D Goldmann, 2022).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Tatiana Avalos-Rendón et al.
The journal of physical chemistry. A, 113(25), 6919-6923 (2009-06-06)
Lithium aluminates (LiAlO(2) and Li(5)AlO(4)) were synthesized, characterized, and tested as possible CO(2) captors. LiAlO(2) did not seem to have good qualities for the CO(2) absorption. On the contrary, Li(5)AlO(4) showed excellent behavior as a possible CO(2) captor. Li(5)AlO(4) was
Wen-Yuan Pang et al.
Recent patents on nanotechnology, 4(1), 32-40 (2010-03-11)
In this paper, we reviewed the current development and patents for the application of high-brightness and high-efficiency white light-emitting diode (LED). The high-efficiency GaN nanostructures, such as disk, pyramid, and rod were grown on LiAlO(2) substrate by plasma-assisted molecular-beam epitaxy
Shan-Li Wang et al.
Journal of hazardous materials, 147(1-2), 205-212 (2007-01-24)
In this study, lithium intercalated gibbsite (LIG) was investigated for its effectiveness at removing phosphate from water and the mechanisms involved. LIG was prepared through intercalating LiCl into gibbsite giving a structure of [LiAl2(OH)6]+ layers with interlayer Cl- and water.
M A Eastman
Journal of magnetic resonance (San Diego, Calif. : 1997), 139(1), 98-108 (1999-07-02)
Hartmann-Hahn match conditions for n2 --> M2 CP/MAS between two quadrupolar nuclei, spin-lock signal as a function of effective nutation frequency, and the correlation of effective nutation frequency and radiofrequency field strength are reported for three samples: sodium diborate (Na2B4O7)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service