Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

347043

Sigma-Aldrich

Lithium peroxide

technical grade, 90%

Synonym(s):

Dilithium peroxide, Lithium oxide (Li2 O2 )

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Li2O2
CAS Number:
Molecular Weight:
45.88
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

90%

form

powder

reaction suitability

reagent type: oxidant

application(s)

battery manufacturing

SMILES string

[Li+].[Li+].[O-][O-]

InChI

1S/2Li.O2/c;;1-2/q2*+1;-2

InChI key

HPGPEWYJWRWDTP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1A

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael J Zachman et al.
Nature, 560(7718), 345-349 (2018-08-17)
Solid-liquid interfaces are important in a range of chemical, physical and biological processes1-4, but are often not fully understood owing to the lack of high-resolution characterization methods that are compatible with both solid and liquid components5. For example, the related
Huan-Feng Wang et al.
Nanoscale, 11(24), 11513-11520 (2019-05-01)
The design of a nanoporous cathode for Li-O2 batteries that can achieve high chemical stability and superior electrochemical performance remains a formidable challenge. A novel approach for the fabrication of nanoengineered PdNi alloys directly coated onto Ni nonwoven fabric (PdNi/NiNF)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service