Skip to Content
MilliporeSigma
All Photos(1)

Documents

307041

Sigma-Aldrich

1-Phenyl-2-pyrrolidinone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11NO
CAS Number:
Molecular Weight:
161.20
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

solid

bp

123 °C/0.2 mmHg (lit.)

mp

67-69 °C (lit.)

SMILES string

O=C1CCCN1c2ccccc2

InChI

1S/C10H11NO/c12-10-7-4-8-11(10)9-5-2-1-3-6-9/h1-3,5-6H,4,7-8H2

InChI key

JMVIVASFFKKFQK-UHFFFAOYSA-N

General description

A novel derivative of 1-phenyl-2-pyrrolidinone, blebbistatin, inhibits non-muscle myocin II activity with high specificity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mark S Duxbury et al.
Biochemical and biophysical research communications, 313(4), 992-997 (2004-01-07)
Blebbistatin is a novel 1-phenyl-2-pyrrolidinone derivative capable of inhibiting non-muscle myosin II activity with a high degree of specificity. We examined the effects of blebbistatin on pancreatic adenocarcinoma cellular migration, invasion, adhesion, and spreading. Blebbistatin dose-dependently inhibited cellular migration and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service