Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

293067

Sigma-Aldrich

Succinonitrile-d4

98 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCD2CD2CN
CAS Number:
Molecular Weight:
84.11
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

form

solid

mp

49-56 °C (lit.)

mass shift

M+4

SMILES string

[2H]C([2H])(C#N)C([2H])([2H])C#N

InChI

1S/C4H4N2/c5-3-1-2-4-6/h1-2H2/i1D2,2D2

InChI key

IAHFWCOBPZCAEA-LNLMKGTHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service