Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

592722

Sigma-Aldrich

4-Aminopyridine-d6

≥98 atom % D, ≥99% (CP)

Synonym(s):

4-Pyridin-2,3,5,6-d4-amine-d2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5D6N2
CAS Number:
Molecular Weight:
100.15
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

≥98 atom % D

Assay

≥99% (CP)

form

solid

bp

273 °C (lit.)

mp

155-158 °C (lit.)

mass shift

M+6

SMILES string

[2H]N([2H])c1c([2H])c([2H])nc([2H])c1[2H]

InChI

1S/C5H6N2/c6-5-1-3-7-4-2-5/h1-4H,(H2,6,7)/i1D,2D,3D,4D/hD2

InChI key

NUKYPUAOHBNCPY-UDDMDDBKSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service