Skip to Content
MilliporeSigma
All Photos(1)

Documents

130621

Sigma-Aldrich

Glutaronitrile

99%

Synonym(s):

1,3-Dicyanopropane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NC(CH2)3CN
CAS Number:
Molecular Weight:
94.11
Beilstein:
1738385
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.434 (lit.)

bp

285-287 °C (lit.)

mp

−29 °C (lit.)

solubility

H2O: soluble
alcohol: soluble
chloroform: soluble

density

0.995 g/mL at 25 °C (lit.)

SMILES string

N#CCCCC#N

InChI

1S/C5H6N2/c6-4-2-1-3-5-7/h1-3H2

InChI key

ZTOMUSMDRMJOTH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Glutaronitrile (1,3-Dicyanopropane) was used in the preparation of dicarbonyl compounds.
Glutaronitrile can be used as:
  • An electrolyte additive in high energy/power Li-ion batteries.
  • A glutarate ligand source to prepare zinc glutarate (ZnGA) by reacting with zinc perchlorate hexahydrate.
  • A reactant to synthesize pentanedithioamide by reacting with Lawesson′s reagent in the presence of boron trifluoride etherate (BF3.OEt2).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xingyong Wang et al.
The Journal of organic chemistry, 78(11), 5273-5281 (2013-05-15)
A palladium-catalyzed addition of potassium aryltrifluoroborates to aliphatic nitriles has been developed, leading to a wide range of alkyl aryl ketones with moderate to excellent yields. Moreover, several dinitriles (e.g., malononitrile, glutaronitrile, and adiponitrile) were applicable to this process for
γ-butyrolactone and glutaronitrile as 5 V electrolyte additive and its electrochemical performance for LiNi0. 5Mn1. 5O4
Xu Yun, et al.
Journal of alloys and compounds, 698, 207-214 (2017)
C Cabezas et al.
Astronomy and astrophysics, 636 (2020-05-28)
Nitriles constitute almost 15% of the molecules observed in the interstellar medium (ISM), surprisingly only two dinitriles have been detected in the ISM so far. The lack of astronomical detections for dinitriles may be partly explained by the absence of
The BF3.OEt2-Assisted Conversion of Nitriles into Thioamides with Lawesson?s Reagent
Nagl Michael, et al.
Synthesis, 2008(24), 4012-4018 (2008)
Hydrothermal synthesis of single-crystalline zinc glutarate and its structural determination
Kim Jong-Seong, et al.
Chemistry of Materials, 16(16), 2981-2983 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service