Skip to Content
MilliporeSigma
All Photos(1)

Documents

291226

Sigma-Aldrich

Diallyl maleate

technical grade, 93%

Synonym(s):

Maleic Acid Diallyl Ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2OCOCH=CHCOOCH2CH=CH2
CAS Number:
Molecular Weight:
196.20
Beilstein:
1725954
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Quality Level

vapor density

6.6 (vs air)

vapor pressure

4 mmHg ( 25 °C)

Assay

93%

refractive index

n20/D 1.469 (lit.)

bp

106-116 °C/4 mmHg

mp

−47 °C (lit.)

density

1.074 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CCOC(=O)\C=C/C(=O)OCC=C

InChI

1S/C10H12O4/c1-3-7-13-9(11)5-6-10(12)14-8-4-2/h3-6H,1-2,7-8H2/b6-5-

InChI key

ZPOLOEWJWXZUSP-WAYWQWQTSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Synthesis and Characterization of Allyl Terpene Maleate Monomer: This study presents a new synthesis route for allyl terpene maleate, differentiating its chemical structure from the typical terpene maleate, which can impact the production of fragrances and flavor industry (Hummel et al., 2019).
  • Platinum-Catalyzed Hydrosilylation in Polymer Chemistry: The inhibitory properties of diallyl maleate in platinum-catalyzed reactions are explored, providing insights into the control of curing processes in polymer manufacturing (Lukin et al., 2020).
  • Polyacrylate Decorating Poly (ethylene terephthalate) Film Surface: A new methyl methacrylate-methyl acrylate-diallyl maleate-maleic acid copolymer was synthesized to enhance the oxygen barrier properties of PET films, which is crucial for packaging applications (Zhong et al., 2021).
  • Preparation and Characterization of DMAA-DAM Gel/Hydrogel: The study focuses on the preparation of a non-aqueous gel using N,N-Dimethyl acrylamide and Diallyl Maleate, aimed at targeted drug release in specific regions of the intestine (Bhuyan et al., 2023).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

249.8 °F - closed cup

Flash Point(C)

121 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service