Skip to Content
MilliporeSigma
All Photos(1)

Documents

291609

Sigma-Aldrich

2,4,6-Triallyloxy-1,3,5-triazine

97%

Synonym(s):

Triallyl cyanurate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15N3O3
CAS Number:
Molecular Weight:
249.27
Beilstein:
235560
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

bp

156 °C

mp

26-27 °C

density

1.11 g/mL at 30 °C

SMILES string

C=CCOc1nc(OCC=C)nc(OCC=C)n1

InChI

1S/C12H15N3O3/c1-4-7-16-10-13-11(17-8-5-2)15-12(14-10)18-9-6-3/h4-6H,1-3,7-9H2

InChI key

BJELTSYBAHKXRW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

347.9 °F

Flash Point(C)

175.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jinujacob George et al.
Nanoscale research letters, 3(12), 508-515 (2008-12-01)
Carbon nanofiber (CNF) is one of the stiffest materials produced commercially, having excellent mechanical, electrical, and thermal properties. The reinforcement of rubbery matrices by CNFs was studied in the case of ethylene vinyl acetate (EVA). The tensile strength was greatly
Jinu Jacob George et al.
Nanoscale research letters, 4(7), 655-664 (2009-01-01)
A series of ethylene vinyl acetate (EVA) nanocomposites using four kinds of EVA with 40, 50, 60, and 70 wt% vinyl acetate (VA) contents and three different carbon-based nanofillers-expanded graphite (EG), multi-walled carbon nanotube (MWCNT), and carbon nanofiber (CNF) have
Fan Zhou et al.
Optics express, 24(6), 6367-6380 (2016-05-04)
Split ring resonators have been studied extensively in reconstituting the diminishing magnetism at high electromagnetic frequencies in nature. However, breakdown in the linear scaling of artificial magnetism is found to occur at the near-infrared frequency mainly due to the increasing
Reka Geczy et al.
Lab on a chip, 19(5), 798-806 (2019-01-29)
Polymeric microfluidic chips offer a number of benefits compared to their glass equivalents, including lower material costs and ease and flexibility of fabrication. However, the main drawback of polymeric materials is often their limited resistance to (organic) solvents. Previously, thiol-ene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service