Skip to Content
MilliporeSigma
All Photos(2)

Documents

269379

Sigma-Aldrich

Diallyl phthalate

97%

Synonym(s):

Allyl phthalate, DAP

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,2-(CO2CH2CH=CH2)2
CAS Number:
Molecular Weight:
246.26
Beilstein:
1880877
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

8.3 (vs air)

vapor pressure

2.3 mmHg ( 150 °C)

Assay

97%

form

liquid

autoignition temp.

725 °F

refractive index

n20/D 1.519 (lit.)

bp

165-167 °C/5 mmHg (lit.)

density

1.121 g/mL at 25 °C (lit.)

SMILES string

C=CCOC(=O)c1ccccc1C(=O)OCC=C

InChI

1S/C14H14O4/c1-3-9-17-13(15)11-7-5-6-8-12(11)14(16)18-10-4-2/h3-8H,1-2,9-10H2

InChI key

QUDWYFHPNIMBFC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

330.8 °F - closed cup

Flash Point(C)

166 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beng H Tan et al.
Advances in colloid and interface science, 113(2-3), 111-120 (2005-06-07)
The effect of cross-linked density on the rheological behavior of model pH-responsive microgel systems consisting of methacrylic acid-ethyl acrylate (MAA-EA) cross-linked with di-allyl phthalate (DAP) was examined. Neutralization of acid groups increases the osmotic pressure exerted by counter-ions trapped in
N Aarab et al.
Aquatic toxicology (Amsterdam, Netherlands), 78 Suppl 1, S86-S92 (2006-04-04)
Environmental pollutants with hormonal activity including bisphenol, diallyl phtalate and tetrabromodiphenyl ether, have the potential to alter gonadal development and reproduction in aquatic wildlife. Little is known about the biological impact of environmentally relevant concentrations in mussels. To investigate some
Jin Zhou et al.
Chemosphere, 79(5), 595-604 (2010-03-02)
The contamination of marine ecosystems by endocrine disrupting compounds (EDCs) is of great concern. Protein expression profile maybe a good method to help us understand the molecular mechanisms of EDCs-toxicity to aquatic organisms. In this study, the abalone (Haliotis diversicolor
Janina Barsiene et al.
Aquatic toxicology (Amsterdam, Netherlands), 78 Suppl 1, S105-S108 (2006-04-18)
Analysis of micronuclei, nuclear buds, bi-polynucleated and fragmented-apoptotic cells was performed in gills of blue mussels exposed for 3 weeks to sublethal concentrations of bisphenol A, diallyl phthalate (for the both nominal concentration 50 ppb) and to tetrabromodiphenyl ether-47 (nominal
A Gomiero et al.
Aquatic toxicology (Amsterdam, Netherlands), 78 Suppl 1, S34-S41 (2006-04-25)
Ciphergen ProteinChip Technology is a proteomic tool, used for the discovery of new and sensitive biomarkers. This approach was used to evaluate the protein profile of crabs exposed to various pollutants. Two different exposure experiments were performed: spider crabs (Hyas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service