Skip to Content
MilliporeSigma
All Photos(5)

Key Documents

289345

Sigma-Aldrich

Cesium fluoride

99.9% trace metals basis

Synonym(s):

NSC 84270

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
CsF
CAS Number:
Molecular Weight:
151.90
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
Assay:
99.9% trace metals basis
Pricing and availability is not currently available.

Quality Level

Assay

99.9% trace metals basis

mp

682 °C (lit.)

density

4.115 g/mL at 25 °C (lit.)

SMILES string

[F-].[Cs+]

InChI

1S/Cs.FH/h;1H/q+1;/p-1

InChI key

XJHCXCQVJFPJIK-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Reactant for:
  • Preparation of building blocks for synthesis of fluoroallylic compounds
  • Synthesis of alcohols via hydrolysis of alkyl silyl ethers at neutral pH in buffered mixed organic-aqueous solutions
  • Nucleophilic fluorination of alkynyliodonium salts to form fluorovinylic compounds
  • Nucleophilic aromatic substitution (SNAr) reactions
Used as a base in a Suzuki cross-coupling synthesis of ortho-substituted biaryls.[1] Also employed as a reagent for nucleophilic fluorination of primary halides and sulfonates in protic media such as tert-butyl and tert-pentyl alcohols.[2]
Used in the successful synthesis of a single-crystal Dion-Jacobson phase, CsLaTa2O7, that has applications in photocatalysis and superconductivity.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Repr. 2 - STOT RE 2

Target Organs

Kidney,Adrenal gland

Supplementary Hazards

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Organometallic Chemistry, 691, 5688-5688 (2006)
Dong Wook Kim et al.
Journal of the American Chemical Society, 128(50), 16394-16397 (2006-12-15)
Aprotic solvents are usually preferred for the SN2 reactions, because nucleophilicity and hence SN2 reactivity are severely retarded by the influence of the partial positive charge of protic solvents. In this work, we introduce a remarkable effect of using tertiary
J Kypr et al.
International journal of biological macromolecules, 13(1), 9-13 (1991-02-01)
Ten DNA fragments containing self-complementary alternating sequences of adenine and thymine differing in length and the starting nucleotide were studied by c.d. spectroscopy. It was found that d(TATATATA) but not d(ATATATAT), d(TATATA), d(CTATATAG) or (dT-dA)20 isomerized into the unusual X-DNA
L Bertollini et al.
Neuroreport, 5(5), 553-556 (1994-01-31)
The effect of intracellular fluoride ions on voltage-dependent calcium currents was tested during whole-cell voltage-clamp recordings in thalamic neurones acutely dissociated from young adult rats. It is demonstrated that 5-30 mM intracellular fluoride selectively and reversibly suppresses the high voltage-activated
S Nelson et al.
Science (New York, N.Y.), 265(5173), 774-777 (1994-08-05)
Neurons in the primary visual cortex of the cat are selectively activated by stimuli with particular orientations. This selectivity can be disrupted by the application of antagonists of the inhibitory neurotransmitter gamma-aminobutyric acid (GABA) to a local region of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service