Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

516988

Sigma-Aldrich

Cesium hydroxide monohydrate

99.95% trace metals basis

Synonym(s):

2-Aminoethyl 2-methylacrylate hydrochloride, 2-Aminoethyl methacrylate hydrochloride, Methacrylic acid, 2-aminoethyl ester, hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCsO · H2O
CAS Number:
Molecular Weight:
167.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.95% trace metals basis
form:
powder

Quality Level

Assay

99.95% trace metals basis

form

powder

impurities

≤550.0 ppm Trace Metal Analysis

mp

272 °C (lit.)

density

3.68 g/mL at 25 °C (lit.)

SMILES string

[Cs+].[O-][H].[H]O[H]

InChI

1S/Cs.H2O/h;1H2/q+1;/p-1

InChI key

HUCVOHYBFXVBRW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cesium hydroxidemonohydrate is commonly used as a catalyst in many organic transformations andas an initiator in polymerization reactions.

Application

Cesium hydroxide monohydrate can be used as a catalyst in the synthesis of:
  • O-allenes and N-allenes via isomerization of terminal alkynes. This method allows the efficient synthesis of allenes in high yields under mild conditions with wide functional group tolerance.
  • Unsymmetrical bis-allyl ethers consist of an E-allylic unit and a terminal allylic unit via dimerization of Baylis-Hillman adducts.
Cesium hydroxide monohydrate can also be used as an initiator for the heterogeneous ring-opening polymerization of monosubstituted oxiranes.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A S Malkani et al.
Science advances, 6(45) (2020-11-08)
Electrolyte cations affect the activity of surface-mediated electrocatalytic reactions; however, understanding the modes of interaction between cations and reaction intermediates remains lacking. We show that larger alkali metal cations (excluding the thickness of the hydration shell) promote the electrochemical CO
David Johannes Scholz et al.
Magma (New York, N.Y.), 28(6), 591-598 (2015-10-10)
(13)C metabolic MRI using hyperpolarized (13)C-bicarbonate enables preclinical detection of pH. To improve signal-to-noise ratio, experimental procedures were refined, and the influence of pH, buffer capacity, temperature, and field strength were investigated. Bicarbonate preparation was investigated. Bicarbonate was prepared and
Andrea Šlampová et al.
Analytica chimica acta, 887, 92-100 (2015-09-01)
Electrolysis is omnipresent in all electrochemical processes including electromembrane extraction (EME). The effects of electrolysis on quantitative aspects of EME were comprehensively evaluated for a set of acidic (substituted phenols) and basic (basic drugs) analytes. EMEs were carried out across

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service