Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

270822

Sigma-Aldrich

Di-tert-butyl acetylenedicarboxylate

98%

Synonym(s):

2-Butynedioic acid di-tert-butyl ester, Di-tert-butyl 2-butynedioate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3COCOC≡CCOOC(CH3)3
CAS Number:
Molecular Weight:
226.27
Beilstein:
1957547
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

80-82 °C/0.05 mmHg (lit.)

mp

33-37 °C (lit.)

SMILES string

CC(C)(C)OC(=O)C#CC(=O)OC(C)(C)C

InChI

1S/C12H18O4/c1-11(2,3)15-9(13)7-8-10(14)16-12(4,5)6/h1-6H3

InChI key

FBCRUXRGQFLOMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The cross-cyclotrimerization of di-tert-butyl acetylenedicarboxylate, silylacetylenes and acrylamides was studied with cationic rhodium(I)/(R)-tol-binap complex as catalyst. Glycosyl azides were subjected to 1,3-dipolar cycloaddition with di-tert-butyl acetylenedicarboxylate.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Hara et al.
Angewandte Chemie (International ed. in English), 53(11), 2956-2959 (2014-02-08)
It has been established that a cationic rhodium(I)/(R)-tol-binap complex catalyzes the cross-cyclotrimerization of silylacetylenes, di-tert-butyl acetylenedicarboxylates, and acrylamides with excellent chemo-, regio-, and enantioselectivities. Unsymmetrical alkynoates can also be employed in place of di-tert-butyl acetylenedicarboxylate for this process, but with
W Bröder et al.
Carbohydrate research, 249(1), 221-241 (1993-10-18)
Glycosyl azides provide reliable anomeric protection stable to conditions for hydrolytic removal of ester groups, for reductive opening or release of acetalic diol protection, for the introduction of ether-type protection, and for glycosylation processes. The utility of this anomeric protection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service