Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

339296

Sigma-Aldrich

Manganese(III) fluoride

99.9% trace metals basis

Synonym(s):

Manganese trifluoride, Manganic fluoride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnF3
CAS Number:
Molecular Weight:
111.93
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

reaction type: C-H Activation
reagent type: catalyst

impurities

≤1500.0 ppm Trace Metal Analysis

density

3.54 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

F[Mn](F)F

InChI

1S/3FH.Mn/h3*1H;/q;;;+3/p-3

InChI key

SRVINXWCFNHIQZ-UHFFFAOYSA-K

Application

Starting material for solid state preparation of Na3MnF6 for further high pressure structural determination studies.

Pictograms

Flame over circleSkull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carlson, S. et al.
Inorganic Chemistry, 37, 1486-1486 (1998)

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service