MilliporeSigma
  • Novel method for simultaneous aqueous in situ derivatization of THC and THC-COOH in human urine samples: validation and application to real samples.
Novel method for simultaneous aqueous in situ derivatization of THC and THC-COOH in human urine samples: validation and application to real samples.
Journal of analytical toxicology (2011-04-26)
S Chericoni, I Battistini, S Dugheri, M Pacenti, M Giusiani
ABSTRACT
The present work describes the validation of a novel aqueous in situ derivatization procedure with trimethyloxonium tetrafluoroborate (TMO) as methylating agent for the simultaneous, quantitative analysis of Δ(9)-tetrahydrocannabinol (THC) and 11-nor-Δ(9)-tetrahydrocannabinol carboxylic acid (THC-COOH) in human urine. The derivatizing agent is directly added to the urine sample and the methyl-derivatives are then recovered by liquid-liquid extraction procedure. Gas chromatography-mass spectrometry was used to detect the derivatives in selected ion monitoring mode. The limits of detection were 0.7 ng/mL for THC and 0.5 ng/mL for THC-COOH, whereas the limits of quantification were 1.9 and 0.9 ng/mL, respectively. The method has been applied to 60 real samples both positive and negative to immunochemical screening test resulting to be very useful and reliable in routine analysis of THC-COOH in human urine for toxicological and forensic purposes.
Sigma-Aldrich
Trimethyloxonium tetrafluoroborate, 95%