Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

270792

Sigma-Aldrich

4-(Chloromethyl)benzoic acid

95%

Synonym(s):

α-Chloro-p-toluic acid, α-Chloro-p-toluylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7ClO2
CAS Number:
Molecular Weight:
170.59
Beilstein:
1907970
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

powder

mp

201-202 °C (lit.)

functional group

carboxylic acid
chloro

SMILES string

OC(=O)c1ccc(CCl)cc1

InChI

1S/C8H7ClO2/c9-5-6-1-3-7(4-2-6)8(10)11/h1-4H,5H2,(H,10,11)

InChI key

OITNBJHJJGMFBN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-(Chloromethyl)benzoic acid was used in the synthesis of dihydroxy stilbene derivatives.

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sheng Yan et al.
Colloids and surfaces. B, Biointerfaces, 188, 110769-110769 (2020-01-10)
Polysulfone (PSf) membrane is widely employed in blood purification fields, but the blood compatibility of PSf membrane is not adequate. To improve the hemocompatibility of PSf membrane, 4-(chloromethyl)benzoic acid (CMBA) and sulfonated hydroxypropyl chitosan (SHPCS) were grafted onto PSf membrane
Marcela Silva Lopes et al.
Anti-cancer agents in medicinal chemistry, 15(2), 206-216 (2014-11-19)
Twenty-seven nitrated and non-nitrated compounds have been synthesized and tested for their growth inhibitory activity on three human cancer cells lines. Fourteen compounds were able to inhibit more than 50% of the growth of at least one of the cancer
Mankil Jung et al.
Bioorganic & medicinal chemistry letters, 17(16), 4481-4486 (2007-06-29)
Dihydroxy stilbene derivatives were designed based on lithospermic acid B and were prepared from 4-(chloromethyl)benzoic acid. The inhibitory activities of the novel compounds against protein tyrosine phosphatase 1B (PTP1B) were evaluated. 3,4-Dihydroxy stilbene carbonyl compounds (7, 11b, 27b) inhibited PTP1B
Yasuo Yoshimi et al.
Analytica chimica acta, 862, 77-85 (2015-02-16)
In this work, the anodic current at an electrode grafted with a molecularly imprinted polymer (MIP) crosslinked via a combination of hydrophobic ethyleneglycol dimethacrylate (EDMA) and hydrophilic methylene bisacrylamide (MBAA) was found to exhibit enantioselective sensitivity to the phenylalanine template

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service