Skip to Content
MilliporeSigma
All Photos(1)

Documents

233315

Sigma-Aldrich

Acetic acid-d4

≥99.9 atom % D

Synonym(s):

Acetic-d3 acid-d, Tetradeuteroacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CD3CO2D
CAS Number:
Molecular Weight:
64.08
Beilstein:
1748971
EC Number:
MDL number:
UNSPSC Code:
12142201
PubChem Substance ID:
NACRES:
NA.21

vapor density

2.07 (vs air)

vapor pressure

11.4 mmHg ( 20 °C)

isotopic purity

≥99.9 atom % D

Assay

≥99% (CP)

form

liquid

autoignition temp.

800 °F

expl. lim.

16 %

technique(s)

NMR: suitable

impurities

≤0.0500% water
water

refractive index

n20/D 1.368 (lit.)

bp

115.5 °C (lit.)

mp

15-16 °C (lit.)

density

1.119 g/mL at 25 °C (lit.)

mass shift

M+4

SMILES string

[2H]OC(=O)C([2H])([2H])[2H]

InChI

1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i1D3/hD

InChI key

QTBSBXVTEAMEQO-GUEYOVJQSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetic acid-d4 is a deuterated NMR solvent that is useful in NMR-based research and analyses.

Application

Acetic acid-d4 may be used in the synthesis of deuterated (7E,9Z)-CLA (conjugated linoleic acid) and Pr(CD3COO)3.D2O (deuterium analog of praseodymium (III) acetate hydrate).

Recommended products

Check out ChemisTwin®, our brand new online portal for identity confirmation and quantification of NMR spectra. Learn more or reach out to us for a free trial.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis and spectroscopic properties of praseodymium (III) acetate hydrate
Hehlen MP, et al
Inorganic Chemistry, 30(10), 2273-2277 (1991)
Reaction of 13N produced by recoil deuterons in pile-irradiated acetic acid-d4 and malonic acid-d4.
Sensui Y, et al.
J. Radioanal. Nucl. Chem., 118(1), 23-31 (1987)
Stereocontrolled synthesis of (7E, 9Z)-[9, 10-2H]-conjugated linoleic acid.
Broustal G and Loreau O.
Journal of Labelled Compounds & Radiopharmaceuticals, 47(12), 875-880 (2004)
Dissociation of Acetic Acid-d4 in Deuterium Oxide from 5 to 50? and Related Isotope Effects.
Paabo M, et al.
The Journal of Physical Chemistry, 70(7), 2073-2077 (1966)
Using high-performance quantitative NMR (HP-qNMR?) for certifying traceable and highly accurate purity values of organic reference materials with uncertainties< 0.1%.
Weber M, et al.
Accreditation and Quality Assurance, 18(2), 91-98 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service