Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

227544

Sigma-Aldrich

trans-2-Ethoxy-5-(1-propenyl)phenol

97%

Synonym(s):

Propenylguaethol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OC6H3(CH=CHCH3)OH
CAS Number:
Molecular Weight:
178.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

86-88 °C (lit.)

SMILES string

CCOc1ccc(\C=C\C)cc1O

InChI

1S/C11H14O2/c1-3-5-9-6-7-11(13-4-2)10(12)8-9/h3,5-8,12H,4H2,1-2H3/b5-3+

InChI key

RADIRXJQODWKGQ-HWKANZROSA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans-2-Ethoxy-5-(1-propenyl)phenol was used in the synthesis of 2S-((E)-2-ethoxy-5-propenyl-phenoxy)-1-(3,4,5-trimethoxy-phenyl)-propane-1-one.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ai-Lan Lee et al.
Organic & biomolecular chemistry, 1(22), 3957-3966 (2003-12-11)
A general asymmetric route to both enantiomers of polysphorin has been developed. The route utilizes polymer-supported reagents, catalysts and scavengers to minimise the need for aqueous work-up and chromatography. This includes application of a method to scavenge 2,3-dichloro-5,6-dicyano-1,4-benzoquinone (DDQ) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service