Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P21951

Sigma-Aldrich

β-Phenylcinnamaldehyde

Synonym(s):

3,3-Diphenylacrolein

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2C=CHCHO
CAS Number:
Molecular Weight:
208.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

bp

195-200 °C/17 mmHg (lit.)

mp

44-46 °C (lit.)

SMILES string

O=C\C=C(\c1ccccc1)c2ccccc2

InChI

1S/C15H12O/c16-12-11-15(13-7-3-1-4-8-13)14-9-5-2-6-10-14/h1-12H

InChI key

MWAFWBDWAWZJGK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service