Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

326364

Sigma-Aldrich

Niobium(IV) chloride tetrahydrofuran complex

Synonym(s):

Tetrachlorobis(tetrahydrofuran)niobium

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NbCl4(CO4H8)2
CAS Number:
Molecular Weight:
378.93
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

form

powder

mp

93 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

Cl[Nb](Cl)(Cl)Cl.C1CCOC1.C2CCOC2

InChI

1S/2C4H8O.4ClH.Nb/c2*1-2-4-5-3-1;;;;;/h2*1-4H2;4*1H;/q;;;;;;+4/p-4

InChI key

OLGCTTOCPNLTKW-UHFFFAOYSA-J

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service