Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

336602

Sigma-Aldrich

Niobium(V) chloride

≥99.9% trace metals basis

Synonym(s):

Niobium pentachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NbCl5
CAS Number:
Molecular Weight:
270.17
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
≥99.9% trace metals basis
form:
crystals and lumps

Quality Level

Assay

≥99.9% trace metals basis

form

crystals and lumps

impurities

≤1,000.0 ppm Trace Metal Analysis

bp

254 °C (lit.)

mp

204.7 °C (lit.)

density

2.75 g/mL at 25 °C (lit.)

SMILES string

Cl[Nb](Cl)(Cl)(Cl)Cl

InChI

1S/5ClH.Nb/h5*1H;/q;;;;;+5/p-5

InChI key

YHBDIEWMOMLKOO-UHFFFAOYSA-I

Looking for similar products? Visit Product Comparison Guide

Application

Used to form mixed chloro-aryloxide compounds, such as [Nb(OC6H3-2,6-i-Pr2)2Cl3]2, which form pyridine or phosphine adducts with a distorted six-coordinate geometry.†

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Janet R. Clark et al.
Inorganic chemistry, 36(17), 3623-3631 (1997-08-13)
The mixed chloro-aryloxide compounds [M(OC(6)H(3)Pr(i)(2)-2,6)(2)Cl(3)](2) (M = Nb (1a), Ta (1b); both structurally characterized) and [M(OC(6)H(3)Pr(i)(2)-2,6)(3)Cl(2)] (M = Nb (2a), Ta (2b)) react with pyridine (py) and PMe(2)Ph to produce a series of adducts cis-mer-[MCl(3)(OC(6)H(3)Pr(i)(2)-2,6)(2)(L)] (M, L: Nb, py (3a);

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service