Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

212830

Sigma-Aldrich

trans-1-Methoxy-3-trimethylsiloxy-1,3-butadiene

95%

Synonym(s):

Danishefsky’s diene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiOC(=CH2)CH=CHOCH3
CAS Number:
Molecular Weight:
172.30
Beilstein:
1616761
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

impurities

2-5% 4-methoxy-3-buten-2-one

refractive index

n20/D 1.454 (lit.)

bp

68-69 °C/14 mmHg (lit.)

density

0.885 g/mL at 25 °C (lit.)

functional group

ether

storage temp.

2-8°C

SMILES string

CO\C=C\C(=C)O[Si](C)(C)C

InChI

1S/C8H16O2Si/c1-8(6-7-9-2)10-11(3,4)5/h6-7H,1H2,2-5H3/b7-6+

InChI key

SHALBPKEGDBVKK-VOTSOKGWSA-N

Looking for similar products? Visit Product Comparison Guide

General description

trans-1-methoxy-3-trimethylsiloxy-1,3-butadiene is a funtionalized Diels-Alder diene. Mukaiyama-Michael-type addition/heterocyclization of trans-1-methoxy-3-trimethylsiloxy-1,3-butadiene (Danishefsky′s diene) with 1,2-diaza-1,3-butadiene has been investigated. Asymmetric hetero-Diels-Alder cyclization of Danishefsky′s diene with benzaldehyde catalyzed by mesoporous inorganic/metalorganic hybrid materials has been reported.

Application

trans-1-Methoxy-3-trimethylsiloxy-1,3-butadiene was used:
  • in the synthesis of sulfone analogues of griseofulvin (sulfogriseofulvins), 4H-1-aminopyrroles and 4,5H-pyrazoles
  • as Diels-Alder diene for the synthesis of pyridones and pyranones
  • as reagent employed in the Mannich-Michael reaction for preparation of piperidinones and enaminones

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Friedrich et al.
Archiv der Pharmazie, 329(7), 361-370 (1996-07-01)
Syntheses of substituted, especially of fluoro substituted benzoxathiole 1,1-dioxides, are described. These derivatives were transformed via the Peterson olefination into substituted 2-alkylidene derivatives 27. Diels-Alder reactions of 27 with 1,1-dimethoxy- and 1-methoxy-3-trimethylsiloxy-1,3-butadiene (30, 32) gave sulfone analogues 31 of griseofulvin
The Journal of Organic Chemistry, 57, 4444-4444 (1992)
Tetrahedron, 49, 397-397 (1993)
Mingji Dai et al.
Journal of the American Chemical Society, 129(3), 645-657 (2007-01-18)
The paper describes the course of cycloadditions of Diels-Alder dienophiles containing linked enyne sites, each substituted with activating groups. Consistently, it was found that in the enyne cases the Diels-Alder reaction occurred specifically at the acetylenic center. Furthermore, it was
Tetrahedron, 49, 1749-1749 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service