Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

8.14551

Sigma-Aldrich

2-(Trimethylsiloxy)-1,3-butadiene

for synthesis

Synonym(s):

2-(Trimethylsiloxy)-1,3-butadiene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiOC4H5
CAS Number:
Molecular Weight:
142.27
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

form

liquid

autoignition temp.

350 °C

bp

124 °C/1013 hPa

transition temp

flash point 12 °C

density

0.82 g/cm3 at 20 °C

storage temp.

2-8°C

SMILES string

[Si](OC(=C)C=C)(C)(C)C

InChI

1S/C7H14OSi/c1-6-7(2)8-9(3,4)5/h6H,1-2H2,3-5H3

InChI key

JOAPBVRQZQYKMS-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 97.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.816 - 0.823
Identity (IR): passes test

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

53.6 °F

Flash Point(C)

12 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service