Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

495956

Sigma-Aldrich

trans-3-(tert-Butyldimethylsilyloxy)-N,N-dimethyl-1,3-butadien-1-amine

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CSi(CH3)2OC(=CH2)CH=CHN(CH3)2
CAS Number:
Molecular Weight:
227.42
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

90%

refractive index

n20/D 1.493 (lit.)

bp

234 °C (lit.)

density

0.878 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[H]\C(=C(\[H])C(=C)O[Si](C)(C)C(C)(C)C)N(C)C

InChI

1S/C12H25NOSi/c1-11(9-10-13(5)6)14-15(7,8)12(2,3)4/h9-10H,1H2,2-8H3/b10-9+

InChI key

NSCKABIMFIKUEF-MDZDMXLPSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

199.4 °F - closed cup

Flash Point(C)

93 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation and Diels-Alder Reactivity of 1-Amino-3-siloxy-1,3-butadienes.
Kozmin SA and Rawal VH.
The Journal of Organic Chemistry, 62(16), 5252-5253 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service