Skip to Content
MilliporeSigma
All Photos(2)

Documents

205176

Sigma-Aldrich

Praseodymium(III,IV) oxide

99.9% trace metals basis

Synonym(s):

Praseodymium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pr6O11
CAS Number:
Molecular Weight:
1021.44
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: praseodymium

density

6.5 g/mL at 25 °C (lit.)

SMILES string

O=[Pr]=O.O=[Pr]=O.O=[Pr]=O.O=[Pr]=O.O=[Pr]O[Pr]=O

InChI

1S/11O.6Pr

InChI key

AICMLAQRDYRMRP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used in the solid state synthesis of a remarkable ternary, reduced molybdenum oxide, Pr4Mo9O18, whose structure contains previously unknown Mo7, Mo13and Mo19 clusters. The new cluster product is a small band gap semiconductor.

Features and Benefits

Used in the solid state synthesis of a remarkable ternary, reduced molybdenum oxide, Pr4Mo9O18, whose structure contains previously unknown Mo7, Mo13and Mo19 clusters. The new cluster product is a small band gap semiconductor.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tortelier, J.; Gougeon, P.
Inorganic Chemistry, 37, 6229-6229 (1998)

Articles

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service