Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

246999

Sigma-Aldrich

Ytterbium(III) oxide

99.9% trace metals basis

Synonym(s):

Ytterbia, Ytterbium oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Yb2O3
CAS Number:
Molecular Weight:
394.08
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: ytterbium

SMILES string

O=[Yb]O[Yb]=O

InChI

1S/3O.2Yb

InChI key

FIXNOXLJNSSSLJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ytterbium(III) oxide (Yb₂O₃) is a wide bandgap(~3.26eV) material with high thermal stability, suitable for optoelectronic devices. It can be a potential host material for development of phosphors and solid-state lighting technologies. Furthermore, Yb₂O₃ nanofibers are being explored for renewable energy applications, particularly in enhancing the efficiency of solar cells and photocatalytic processes.

Application

  • Ytterbium (III) oxide reinforced novel TeO2–B2O3–V2O5 glass system: Investigation of the optical, structural, physical, and thermal properties of ytterbium oxide reinforced glass, highlighting improvements in material performance (Kilic et al., 2021).
  • Sonochemical preparation of a ytterbium oxide/reduced graphene oxide nanocomposite for supercapacitors with enhanced capacitive performance: Discusses the synthesis and application of ytterbium oxide/graphene nanocomposites in energy storage devices (Naderi et al., 2016).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joseph K Swabeck et al.
Journal of the American Chemical Society, 140(29), 9120-9126 (2018-06-21)
Semiconductor quantum dot (QD)-sensitized lanthanide ions hold great promise in producing a broadly absorbing and sharply emitting luminophore, but their synthesis has proven to be difficult. We report the first synthesis of core/shell/shell InP/Ln xY1- xF3/ShF3 (Ln = Yb, Nd;
Zhengze Yu et al.
ACS nano, 9(11), 11064-11074 (2015-10-13)
Photodynamic therapy (PDT) is a well-established modality for cancer therapy, which locally kills cancer cells when light irradiates a photosensitizer. However, conventional PDT is often limited by the extremely short lifespan and severely limited diffusion distance of reactive oxygen species

Articles

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Rechargeable solid-state batteries are becoming increasingly important due to wide-spread use in computers, portable electronics, and vehicular applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service