Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

188573

Sigma-Aldrich

5-Methyl-1,3,4-thiadiazole-2-thiol

99%

Synonym(s):

2-Mercapto-5-methyl-1,3,4-thiadiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4N2S2
CAS Number:
Molecular Weight:
132.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

188-189 °C (lit.)

solubility

chloroform: soluble 10 mg/mL, clear to very slightly hazy, colorless

SMILES string

Cc1nnc(S)s1

InChI

1S/C3H4N2S2/c1-2-4-5-3(6)7-2/h1H3,(H,5,6)

InChI key

FPVUWZFFEGYCGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

5-Methyl-1,3,4-thiadiazole-2-thiol is a pharmaceutical intermediate. UV and UV/H2O2 induced degradation of 5-methyl-1,3,4-thiadiazole-2-thiol has been investigated.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonio Lopez et al.
Annali di chimica, 92(1-2), 41-51 (2002-05-08)
The degradation of 5-methyl-1,3,4-thiadiazole-2-thiol (MMTD), a pharmaceutical intermediate found in some aquifers of Northern Italy, has been investigated by means of UV and UV/H2O2 treatments. The study has been carried out with a 17 W low pressure mercury lamp at

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service