Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

530654

Sigma-Aldrich

5-Methylthio-1,3,4-thiadiazole-2-thiol

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4N2S3
CAS Number:
Molecular Weight:
164.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

mp

138-141 °C (lit.)

SMILES string

CSc1nnc(S)s1

InChI

1S/C3H4N2S3/c1-7-3-5-4-2(6)8-3/h1H3,(H,4,6)

InChI key

CFWGYKRJMYXYND-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electron donors and cyanoimidazole acceptors: cyclic voltammetry and molecular orbital study.
Electron donors and cyanoimidazole acceptors: cyclic voltammetry and molecular orbital study.
Allan DS, et al.
Synt. Metals, 25(2), 139-155 (1988)
Adsorption and trace detection of pharmacologically significant 5-methylthio-1, 3, 4-thiadiazole-2-thiol molecule adsorbed on silver nanocolloids and understanding the role of Albrecht's ?A? and Herzberg-Teller contributions in the SERS spectra.
Chowdhury J, et al.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 135, 935-946 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service